Would you like to log in again?
Single Pulley Bench Mount
Supplier: EISCO SCIENTIFIC LLC
PH0290
470230-096EA 15.2 USD InStock
470230-096
Single Pulley Bench Mount
Educational Materials Physics Educational Materials Mechanics Learning Activities
Bench mounted aluminum pulley.
  • Single pulley
  • Bench mounted
  • Aluminum
  • 50 mm dia.
  • For clamping to benches/boards up to 28 mm thick
Bench mounting, aluminum 50 mm diameter, in metal frame for clamping to benches or boards up to 28 mm thick.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty