Would you like to log in again?
Pulley with Universal Clamp
Supplier: EISCO SCIENTIFIC LLC
PH0293
470230-098EA 40.45 USD InStock
470230-098
Pulley with Universal Clamp
Educational Materials Physics Educational Materials Mechanics Learning Activities
Pulley attached to universal clamp.
  • Clamped onto edge up to 1.5" thick
  • 70 mm dia. pulley
Consists of a 70 mm diameter light alloy pulley, held in alloy bracket, with two clamping screws for bench and rod support. The bracket may be clamped to a bench edge or similar support up to 1.5" thick, screwed to a flat surface, or held on 13 mm diameter support rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty