Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Plastic Pulley with Clamp

Supplier: EISCO SCIENTIFIC LLC
PH0296B
470230-102EA 24.8 USD InStock
470230-102
Plastic Pulley with Clamp
Educational Materials Physics Educational Materials Mechanics Learning Activities
Plastic pulley with two- part aluminum frame.

  • 50 mm dia. nylon ball bearing pulley
  • Two-part aluminum frame
  • Overall length of frame 110 mm
  • Maximum clamp opening 25 mm

Comprising a 50 mm diameter nylon ball bearing pulley, carried on a two part aluminum frame, designed primarily for clamping to the edge of a bench, table etc. With the pulley overhanging the edge and in a vertical plane. Overall length of frame (pulley bar) 110 mm. Maximum clamp opening 25 mm.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty