Would you like to log in again?
Pulley with Adjustable Clamp
Supplier: EISCO SCIENTIFIC LLC
PH0297A
470230-104EA 29.75 USD InStock
470230-104
Pulley with Adjustable Clamp
Educational Materials Physics Educational Materials Mechanics Learning Activities
Metal pulley with rotatable pulley for experiment set up.
  • Sturdy metal construction throughout
  • Ideal for demonstration of wide variety of mechanics principles
  • 2" (5 cm) dia. sheave
  • Completely rotatable pulley for flexibility in experiment setups
  • Adjustable to 9" height with 1.9" maximum clamp opening
Sturdy, metal clamp pulley can be used to demonstrate a wide variety of mechanics principles. The metal sheave is 5 cm in diameter and can be adjusted from flush with the tabletop to 13 cm above it. Pulley can be rotated to 360º to suit any experimental setup.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty