Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Force Table Pulley

Supplier: EISCO SCIENTIFIC LLC
PH0300C
470230-108EA 14.8 USD InStock
470230-108
Force Table Pulley
Educational Materials Physics Educational Materials Mechanics Learning Activities
Plastic pulley with table clamp, includes hand knob for tightening.

  • Plastic 2" pulley
  • Clamp made of hard plastic
  • Clamp max opening 1.250"
  • Grey hand knob for tightening clamp

labs Low Friction Force table 2" Pulley - Clamp max opening is 1.25" Clamp is made of hard plastic. Grey hand knob for tighten clamp

Ordering information: The corresponding force table (470014-356) is also available for purchase.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty