Would you like to log in again?
Hooke's Law Apparatus with Weights
Supplier: EISCO SCIENTIFIC LLC
PH0323A
470230-118EA 40.85 USD InStock
470230-118
Hooke's Law Apparatus with Weights
Educational Materials Physics Educational Materials Mechanics Learning Activities
Hooke's Law apparatus, includes weights for demonstration.
  • 15 cm adjustable scale
  • Scale marked in mm
  • Includes experimental notes
  • Wooden base
This complete apparatus has a 15cm adjustable scale. The adjustable scale helps to prevent parallax error. Scale is marked in millimeters and mounted on sturdy 30 cm support rod and a wooden base. A hook supporting a coiled spring with a mass hanger and indicator is attached to rod. 10 g weight plates included along with experimental notes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty