Would you like to log in again?
Loop the Loop with Scale
Supplier: EISCO SCIENTIFIC LLC
PH0323LS
470230-120EA 81.65 USD InStock
470230-120
Loop the Loop with Scale
Educational Materials Physics Educational Materials Mechanics Learning Activities
Loop the Loop apparatus to demonstrate potential to kinetic energy.
  • To demonstrate transformation of potential into kinetic energy
  • For demonstration of conservation of energy, momentum and projectile motion
  • Total Height - Approx. 21”
  • Base Size 23” x 6” Base
  • Includes 0.75” steel ball
  • Loop the Loop looks like the track of a roller coaster
Labs Deluxe Loop Apparatus with scale. To demonstrate transformation of potential into kinetic energy. For demonstration of conservation of energy, momentum and projectile motion. Loop the Loop looks like the track of a roller coaster. The curved metal V track allows you to drop a steel ball from the top and watch the results. Depending on the height at which it is dropped the ball will roll completely around the interior of the track or will drop to the ground. Track has yellow painted scale A circular scale is attached to the device for measuring the angle at which the ball drops and the track is also graduated to determine the distance. Includes 0.75" steel ball. Base Size 23" x 6" Base Total Height - Approx. 21"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty