Would you like to log in again?
Inclined Plane Aluminum
Supplier: EISCO SCIENTIFIC LLC
PH0339
470230-128EA 113.4 USD InStock
470230-128
Inclined Plane Aluminum
Educational Materials Physics Educational Materials Mechanics Learning Activities
Aluminum inclined plane with protractor.
  • Solid aluminum construction
  • Plane length of 480 x 70 mm
  • Includes protractor for angle
This new Inclined plane has a solid aluminum construction, comprises of a pulley which can be adjusted. The length of the plane is 480 x 70 mm approx. and has a protractor for angle measurements. A very useful apparatus to Investigate acceleration, friction, gravity, Galileo's free-fall experiments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty