Would you like to log in again?
Hall’s Carriage
Supplier: EISCO SCIENTIFIC LLC
PH0343A
470230-130EA 37.7 USD InStock
470230-130
Hall’s Carriage
Educational Materials Physics Educational Materials Mechanics Learning Activities
Plastic carriage for motion experiments.
  • Carriage made of plastic resin
  • Wheels of carriage made of aluminum
  • Area in carriage for placement of various weights
Hall's carriage is a great tool that is useful for teaching students about dynamics when used in conjunction with an incline plane. The carriage has a compartment in which different weights can be placed to measure the effect of weight on speed and acceleration. It provides an effective method for students to learn both visually and kinesthetically about work, power, and energy.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty