Would you like to log in again?
Force Board Kit
Supplier: EISCO SCIENTIFIC LLC
PH0345
470230-136EA 116 USD InStock
470230-136
Force Board Kit
Educational Materials Physics Educational Materials Mechanics Learning Activities
Kit to demonstrate force experiments.
  • Force board grid has a coarse resolution of 5 x 5cm and a fine resolution of 1 × 1 cm
  • Clamp on pulleys are constructed of ABS plastic with low friction ball bearings
  • More than enough masses and string to do any arrangement of vectors and forces
A great free standing device to introduce forces and vectors to your classroom. High contrast grid made it force configurations easy to see from any angle in class
Ordering information: Kit contains: (2) 1.5" Low Friction Single pulleys, (4) S Hooks, (1) Roll of String, (20) 10g Masses, (1) 55 cm x 37.5 cm Backboard with feet
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty