Would you like to log in again?
Collision in Two Dimensions Apparatus
Supplier: EISCO SCIENTIFIC LLC
PH0349CTD
470230-146EA 30.95 USD InStock
470230-146
Collision in Two Dimensions Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
Collison in two dimensions apparatus with accessories included.
  • Consists of 25cm one piece track, 3 balls (12 mm dia.), 2 steel, 1 glass, and 1 plumb bob
  • Requires a clamp to attach to end of table (not included)
This apparatus consists of a curved track with a base at one end. On the base is a support to hold a ball at the proper height for a center to center collision with a second ball rolling down the track. The track is level so collision occurs only in the horizontal plane, simplifying calculations. Consists of 25 cm one piece track, 3 balls 12 mm diameter, 2 steel, 1 glass and 1 plumb bob.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty