Would you like to log in again?
Double Cone and Plane
Supplier: EISCO SCIENTIFIC LLC
PH0349DCP
470230-148EA 41.9 USD InStock
470230-148
Double Cone and Plane
Educational Materials Physics Educational Materials Mechanics Learning Activities
Demonstrates center of mass with wooden plane and double cone that comes assembled.
  • Center of mass demonstration
  • Apparatus comprises of a wooden A from with diverging rails and a double ended cone
  • Easy use, place cone of the bottom of the ramp and let go
  • Constructed of durable wood and clear coated for decades of use
  • Comes assembled
This is a great toy, executive gift, or physics demonstration as the cone appears to be traveling "Up" the ramp. The physics behind the demonstration is that as the double-ended cone moves up the ramp the center of mass is actually moving downward which drives the cone up the ramp itself. Amaze your students and colleagues.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty