Would you like to log in again?
Hall's Car Single
Supplier: EISCO SCIENTIFIC LLC
PH0351HCS
470230-154EA 13.65 USD InStock
470230-154
Hall's Car Single
Educational Materials Physics Educational Materials Mechanics Learning Activities
Plastic hall's car used for dynamics experiments.
  • Hall's car
  • Molded one-piece body with a central well to hold additional masses
  • Hole in one end of the body to attach cords
Can be used with an inclined plane, weights, and pulleys to study relationship between energy and work. It features a molded one-piece body with a central well to hold additional masses. The molded wheels have sleeve bearings. There is a hole in one end of the body to attach cords.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty