Would you like to log in again?
Conductometer, 6 Rod
Supplier: EISCO SCIENTIFIC LLC
PH0377
470230-160EA 37.05 USD InStock
470230-160
Conductometer, 6 Rod
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Conductometer with variety of metals to demonstrate heat conductance by various metals.
  • Conductometer is a total of 42 cm long with 9 cm long sample bars
  • Bars are made of zinc, aluminum, brass, copper, stainless steel, iron
  • Place wax in the end of each bar or place a temperature sensor on each bar and heat the center
  • Recommended for use with the Neulog NUL-203 temperature sensor and bunsen burner CH0993NG
Different metals conduct heat at different rates due to their atomic structure. With this demonstration students either clip temperature sensors to each metal bar, or place wax in the end of the bar to see which bar heats up the quickest when the center of the apparatus is held over and open flame
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty