Would you like to log in again?
Radiometer Economy
Supplier: EISCO SCIENTIFIC LLC
PH0418A
470230-174EA 18.45 USD InStock
470230-174
Radiometer Economy
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Radiometer with four lightweight metal arms.
  • Includes black and white vanes in color
  • Mounted on round plastic molded base
  • Glass bulb with 70 mm dia.
Comprising partly evacuated glass bulb approx. 70 mm diameter, containing at its center a fine pivot which supports four light weight metal arms. One side of each vane is blackened, the reverse side is bright. Mounted on round plastic molded base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty