Would you like to log in again?
Thermocouple Copper Constantan
Supplier: EISCO SCIENTIFIC LLC
PH0426
470230-176EA 19.95 USD InStock
470230-176
Thermocouple Copper Constantan
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Simple Thermocouple for demonstrating wide range of temperatures.
  • Type T thermocouple
  • Copper and Constantan
  • Great repeatability at lower temperatures
  • Well suited to oxidizing atmospheres
Very simple T-Type thermocouple comprised of copper and constantan. Use electricity to measure temperature. Great for demonstrations in the lab or classroom or for simply measuring a wide range of temperatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty