Would you like to log in again?
Specific Heat Cylinders with Wood Base
Supplier: EISCO SCIENTIFIC LLC
PH0432
470230-178EA 61.85 USD InStock
470230-178
Specific Heat Cylinders with Wood Base
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Metal cylinder set for demonstrating density investigation and specific heat experiments.
  • Perfect for use in comparing densities of metals or specific heats
  • All cylinders are 16 mm in dia., 100 g (+/- 0.6g), and are drilled for suspension
  • Metals included: Lead, Copper, Brass, Iron, Zinc, Aluminum (from most to least dense, respectively)
  • Finished wooden base 6 7/8" x 1 3/4" x 1 3/4" perfect for storage and display
This cylinder set is perfect for laboratory use to measure and calculate density and/or specific heat. The set includes the following metals: Lead - Density = 11.34g/cm3, Height = 1 13/16 Copper - Density = 8.96g/cm3, Height = 2 1/4 Brass - Density = 8.73g/cm3, Height = 2 3/8 Iron - Density = 7.87g/cm3, Height = 2 5/8 Zinc - Density = 7.14g/cm3, Height = 2 15/16 Aluminum - Density = 2.7g/cm3, Height = 7 5/16.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty