Would you like to log in again?
Equal Mass Density Set
Supplier: EISCO SCIENTIFIC LLC
PH0432CB
470230-180EA 21.85 USD InStock
470230-180
Equal Mass Density Set
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Cylinder set for demonstrating density investigation and specific heat experiments.
  • All cylinders have same diameter to show principles of mass and density between various materials
  • Acrylic, nylon, PVC, aluminum, and brass cylinders
Equal mass density cylinder set of different materials. 5 cylinders included: Acrylic, Nylon, PVC, Aluminum, and Brass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty