Would you like to log in again?
Specific Heat Cylinders Set
Supplier: EISCO SCIENTIFIC LLC
PH0434
470230-182EA 45.25 USD InStock
470230-182
Specific Heat Cylinders Set
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Metal cylinder set for demonstrating density investigation and specific heat experiments.
  • Specific heat cylinders in beautiful wooden storage block
  • Six pieces: copper, lead, brass, zinc, iron, aluminum
  • 38x9.5mm dia. cylinders in a 5" x 1.25" x 1" storage block
  • Use to study the measurable amount of heat required to alter an objects temperature
  • Cylinders have holes drilled in them for easy hands free use
This beautifully crafted set contains six cylinders, 38x9.5mm each. The set is comprised of lead, copper, brass, zinc, iron and aluminum and comes in a premium wooden block for storage and display. Great for the classroom and home. Use for the study of specific heat across a spectrum of objects by determining the amount of heat required to elevate the temperature of an object's mass by 1 Kelvin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty