Would you like to log in again?
Hero's Engine
Supplier: EISCO SCIENTIFIC LLC
PH0498A
470230-192EA 39.1 USD InStock
470230-192
Hero's Engine
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Functional glass model of steam turbine.
  • A functional glass model of the earliest form of steam turbine. Comprising a borosilicate glass bulb 70 mm dia. approx.
  • Comprising a borosilicate glass bulb 70 mm dia. approx., with side arms bent at right angles and formed into jets at their tips
  • The bulb is carried upon a metal stand which leaves it free to rotate about its horizontal axis. If about 25 to 30 ml of water introduced into the bulb and boiled the issuing steam causes rapid rotation
  • 10" tall
  • Clearance under the bulb is 5" to 6.5" for the Bunsen burner
A functional glass model of the earliest form of steam turbine. Comprising a borosilicate glass bulb 70 mm diameter approx., with side arms bent at right angles and formed into jets at their tips. The bulb is carried upon a metal stand which leaves it free to rotate about its horizontal axis. If about 25 to 30 ml of water introduced into the bulb and boiled the issuing steam causes rapid rotation. Supplied complete on stand but without burner. Clearance under the bulb is 5" to 6.5" for the Bunsen burner Please exercise caution while using this unit, as it involves flames, hot vapors and glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty