Would you like to log in again?
Plane Mirrors, Glass Mounted
Supplier: EISCO SCIENTIFIC LLC
PH0515B
470230-196EA 4.69 USD InStock
470230-196
Plane Mirrors, Glass Mounted
Educational Materials Physics Educational Materials Optics Learning Activities
Plane mirrors for studying optics.
  • Silver-backed glass mirror mounted on wooden block measuring approx. 3 x 2 x 0.75" (75 x 50 x 20 mm)
  • Wood mount protects mirror and makes transport simple
  • Plane mirror reflects light at the angle of incidence
  • Small size allows mirror to be passed out and around to a class easily
  • Useful for demonstrating the physics of light and reflection, and can be used in optics setups
This wood-mounted plane mirror is perfect for demonstrating light and reflection physics! Light hitting the mirror will reflect at the same angle. Try it with a flashlight or laser pointer. The mirror's small size make it good for classroom demonstrations, or for use as a spot mirror.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty