Would you like to log in again?
Spectrometer Advanced
Supplier: EISCO SCIENTIFIC LLC
PH0620
470230-220EA 913.9 USD InStock
470230-220
Spectrometer Advanced
Educational Materials Physics Educational Materials Optics Learning Activities
Spectrometer for studying concepts of optics.
  • Includes: Spectrometer, 1 prism holder, and 1 grating holder
  • Solid base with turntable, prism table, collimator, and telescope
  • Used to determine wavelength of light
  • Used to determine the index of refraction of solids and liquids
  • Stored in polished wooden case
The advanced spectrometer, including 1 prism holder and 1 grating holder has a solid base with a turntable, prism table, collimator, and telescope, is ideal for determining both the wavelength of light and the index of refraction of solids and liquids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty