Would you like to log in again?
Colored Filters, Unmounted, Set 6
Supplier: EISCO SCIENTIFIC LLC
PH0645
470230-226EA 18.15 USD InStock
470230-226
Colored Filters, Unmounted, Set 6
Educational Materials Physics Educational Materials Optics Learning Activities
Primary colors filter set.
  • 3 primary colors included: Red, Yellow, Blue.
  • 3 secondary colors included: Green, Orange, Violet
  • Great for color mixing and filtering
  • Made of plastic
  • Squares are 10 x 10 cm
6 piece color filter set, unmounted 10cm squares, 3 primary colors, 3 secondary colors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty