Would you like to log in again?
Wave Form Helix, 10 cm Closed Height
Supplier: EISCO SCIENTIFIC LLC
PH0708A
470230-238EA 18.25 USD InStock
470230-238
Wave Form Helix, 10 cm Closed Height
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Closely wound steel helix for studying wave behavior.
  • Wave form helix
  • 7.5 cm diameter
  • 10 cm long when closed
  • Up to 3m in length when fully stretched
This is a Wave Form Helix that can be: 1) used as a scientific instrument to measure a wave function (it actually has a higher spring constant than the classic toy); or 2) unleashed down a wicked set of stairs. It has a 7.5 cm diameter, 10 cm long when closed, and up to 3 meters in length when fully stretched.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty