Would you like to log in again?
Stroboscope
Supplier: EISCO SCIENTIFIC LLC
PH0752A
470230-254EA 29.75 USD InStock
470230-254
Stroboscope
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Sturdy stroboscope ideal for studying periodic motion.
  • 9.5 inch diameter rotating disc on handle
  • Total height: 18 inches
  • Sturdy plastic disc with slots
  • Disc includes finger hold
  • Handle with rubber grip
The sturdy stroboscope with marked disc, finger hold in disc, and rubber gripped handle is easy to use and ideal for studying periodic motion and speed of rotation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty