Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Synchronized Digital Ripple Wave Generator

Supplier: EISCO SCIENTIFIC LLC
PH0769B
470230-262EA 254.75 USD InStock
470230-262
Synchronized Digital Ripple Wave Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Stroboscope light synchronizes with ripple generator.

  • For use with ripple tanks in classroom demonstration and physics experiments
  • Synchronizes frequency of stroboscopic flashes with the generated water ripples in the tank
  • Attaches to strobe light and vibration generator via 4 mm leads
  • 120VAC, 50/60Hz

This Digital Ripple Wave Generator is designed for used with ripple tanks. Unit connects to both a stroboscopic light and a vibration generator via 4mm plugs. This is a very unique apparatus which provides a stroboscopic light with pulses to synchronize with a ripple generator. Both these parameters function in fully synchronized fashion to generate the stable and visible ripple pattern. The motion frequency / ripple frequency and their amplitudes are controlled electronically. Frequency range is from 0.5 Hz to 200 Hz. Uses 120 V AC, 50/60 Hz power.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty