Would you like to log in again?
Demonstration Electromagnet
Supplier: EISCO SCIENTIFIC LLC
PH0801DE
470230-268EA 34.95 USD InStock
470230-268
Demonstration Electromagnet
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Small electromagnet to demonstrate the concept of electromagnetism.
  • Mounted on plastic case with 4 mm safety connections
  • Maximum of 6 V
  • Paper clips are ideal to use as weights to demonstrate this
A small electromagnet mounted on a plastic case with 4mm safety connections. The voltage supplied through those connections power the electromagnet (maximum of 6V). Students can observe the change in strength of the electromagnet by changing the input voltage. Paper clips are ideal to use as weights to demonstrate this.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty