Would you like to log in again?
Solenoid, Small
Supplier: EISCO SCIENTIFIC LLC
PH0804D
470230-278EA 126 USD InStock
470230-278
Solenoid, Small
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Durable solenoid for induction experiments.
  • 700 turns
  • Insulated Copper
  • 1.6 amps continuous current
  • 5 amps intermittent current
  • Air Cored 1.77 inch dia., 5.9 inch length
Labs Small Solenoid is wound with 700 turns of insulated copper wire and designed to carry 1.6 amps continuously or 5 amps intermittently. It is excellent for studying electricity and magnetism in experiments requiring an external magnetic field. It has an inner diameter of 1.77 inches and length of 5.9 inches allowing for insertion of an experimental apparatus into its magnetic field.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty