Would you like to log in again?
Magnetic Field Chamber, Eisco Scientific
Supplier: EISCO SCIENTIFIC LLC
PH0805
470230-282EA 178.45 USD InStock
470230-282
Magnetic Field Chamber, Eisco Scientific
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Visualize magnetic fields in three dimensions.
  • Demonstrations or to pass around the class for hands on experience
  • Mess free design
This large magnetic field chamber is great for the classroom. It allows students to visualize magnetic fields in three dimensions, instead of the usual two dimensions of most iron filings field visualizers. Perfect for demonstrations and individual hands on experience. The Iron filings are sealed inside the chamber so there are no loose filings to clean up. Simply insert a bar magnet (not included) into the middle of the chamber to view 3-D magnetic field lines.
Ordering information: Magnet not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty