Would you like to log in again?
Demonstration Induction Coil
Supplier: EISCO SCIENTIFIC LLC
PH0840
470230-296EA 51.2 USD InStock
470230-296
Demonstration Induction Coil
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
High quality demonstration induction coil.
  • Both coils fitted with 4 mm sockets for easy use with standard banana plugs
  • Magnetic iron core is 160 mm in length and 15 mm in dia.
  • RoHS compliant and built to last
Primary coil constructed of enameled copper wire, approximate resistance 0.5 Ohms wound on tube with length of 110 x 30 mm in diameter. Secondary coil fits outside primary coil. Coil is constructed of fine enameled copper wire with an approximate resistance of 100 Ohms. Coil is wound on a cylinder with the dimensions 120 mm in length x 82 mm in diameter.
This high quality demonstration induction coil(s) are perfect for showing Faraday's law or incorporating into a bench top research experiment. The set includes 2 nesting coils and cylindrical bar magnet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty