Would you like to log in again?
Eddy Current Demonstration
Supplier: EISCO SCIENTIFIC LLC
PH0845EDU
470230-300EA 37.35 USD InStock
470230-300
Eddy Current Demonstration
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Eddy current for demonstrating Lenz's law.
  • Labs 13" eddy current demonstration
  • Heavy duty 13" copper tube
  • Includes 2 masses, one is steel, one is a neodymium magnet
  • These currents oppose downward motion through a moving magnetic field created by the falling neodymium magnet
Labs 13 eddy current demonstration. This demonstrates the amazing Lenz's law. Consists two masses drop down a 33cm copper tube. One is steel, one is a neodymium magnet. Two masses drop down a 33cm copper tube. One is steel, one is a neodymium magnet. When students see the magnetic mass falls more slowly than the steel mass, you can introduce them to electrical eddy currents. These currents oppose downward motion through a moving magnetic field created by the falling neodymium magnet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty