Would you like to log in again?
Leyden Jar
Supplier: EISCO SCIENTIFIC LLC
PH0906
470230-314EA 59.4 USD InStock
470230-314
Leyden Jar
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Leyden jars for static electricity activities.
  • Useful for performing many different activities in static electricity
  • Comes with inner and outer cup, as well as plastic dielectric
Shock your students again and again with this Leyden Jar. The first device capable of storing an electric charge was the Leyden Jar. Invented by a German, Ewald G. von Kleist, on November 4, 1745. Kliest made the discovery by accident. While experimenting with electricity, he touched his electric generator to a nail stuck into a medicine bottle through the cork. Later, he received a great shock by touching the nail. Although he did not understand how it worked, he had discovered that the nail and the jar were capable of temporarily storing electrons. Today we would call this device a capacitor. Capacitors are used in every form of electronic equipment. This Leyden Jar is designed for classroom demonstrations, and comes with a durable inner and outer plate, a plastic dielectric, and instructions. The inner plate has a hook and ball attached which makes it easy to charge. Use this equipment to duplicate many classic static electricity experiments. Learn about static electricity and the function of a dielectric with this unique device. Leyden Jar is 3" in diameter and 8" tall.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty