Would you like to log in again?
Electroscope Flask with Aluminum Leaf
Supplier: EISCO SCIENTIFIC LLC
PH0912
470230-316EA 17.8 USD InStock
470230-316
Electroscope Flask with Aluminum Leaf
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrates observation of polarity.
  • 7.5" height with 3.5" dia.
  • Glass conical flask
  • Brass rod and aluminum leaf
  • Detect electric charges and determine polarity
Aluminum leaf electroscope used to observe polarity through electric charge detection. It is comprised of a glass conical flask with a brass rod, disc and aluminum leaf supported by a rubber cork.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty