Would you like to log in again?
Needle Electroscope
Supplier: EISCO SCIENTIFIC LLC
PH0917
470230-318EA 35.2 USD InStock
470230-318
Needle Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for experiments with electrostatic principles.
  • Great tool to discover electrostatic principles such as conduction, induction and coulomb repulsion
  • Clear front pane lifts up to allow easy application of ionizing materials
  • Perfect for hands on experience in the classroom
This electroscope is great for hands on experience with electrostatic principles in physics classrooms and labs. The sheet metal walls of the electroscope are attached to a plastic base and top. Front glass window slides up to allow insertion of ionizing material in chamber. A light-weight metal vane is used as a pointer in the electroscope. This light weight metal blade deflects when the electroscope plate is charged. A graduated scale is provided to measure the angle of the deflection. Fitted with 4mm socket. Assembled, the electroscope measures 4 x 3.75 x 2.25 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty