Would you like to log in again?
Demonstration Electroscope
Supplier: EISCO SCIENTIFIC LLC
PH0917A
470230-320EA 46.35 USD InStock
470230-320
Demonstration Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrates static potentials.
  • 150 mm diameter ring
For the demonstration of static potentials. Available with charging ball plus a set of condenser plates with insulated rod. Ring diameter 150 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty