Would you like to log in again?
Van de Graaff Generator Hand Crank
Supplier: EISCO SCIENTIFIC LLC
PH0919A
470230-324EA 378 USD InStock
470230-324
Van de Graaff Generator Hand Crank
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal electrostatic tool for generating voltages.
  • Includes generator and accessories kit
  • Hand operated
  • Metal Sphere
  • Smooth finish wood base
  • Height (with sphere attached): Approx. 24"
The Van de Graaff generator apparatus is an ideal electrostatic tool with which to generate voltages. The included accessories kit provides multiple tools with which to demonstrate the effects of electrostatically produced voltage. This tool provides a visually and kinesthetically effective method for studying the electrostatic behavior of particles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty