Would you like to log in again?
Van de Graaff, 200 kV
Supplier: EISCO SCIENTIFIC LLC
PH0920A
470230-326EA 320.85 USD InStock
470230-326
Van de Graaff, 200 kV
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal electrostatic tool for generating voltages.
  • Up to 200 kV depending on conditions with up to a 60 mm spark
  • Uses 110 or 220 V AC
  • 150 mm diameter sphere
  • Acrylic shaft allows for full visibility
Specially designed for electrostatic experiments and where continuous source of high voltage is required. Fitted on base with smooth running AC Motor, operates on 110 or 220 volts AC. Charge collecting Belt: Silicone rubber having excellent insulation resistance. Tracking: Belt-tracking easily adjusted. Charge collecting combs: Aluminum mesh, clearly visible. Spheres : Aluminum 150 mm dia. Connections : 4mm sockets in dome and base. Voltage developed: Up to 200 kV, depending on ambient conditions with a spark length of 60 mm. Discharger: Spherical 100 mm dia., with insulated handle and 4mm sockets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty