Would you like to log in again?
Electrodes
Supplier: EISCO SCIENTIFIC LLC
PH0927H PH0927J PH0927A PH0927E PH0927G PH0927F PH0927B PH0927I PH0927D
470230-338EA 3.75 USD InStock
470230-338 470230-336 470230-346 470230-334 470230-344 470230-332 470230-342 470230-330 470230-340
Electrodes
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
High quality electrode strip.
  • Very high quality electrode strips
  • 100 x 19 mm size
Aluminum Electrodes Strips - Used for student's electrochemical experiments , very high quality electrodes strips - Has stamped material identifier on strip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty