Would you like to log in again?
Battery Holder
Supplier: EISCO SCIENTIFIC LLC
PH0957D
470230-352EA 39 USD InStock
470230-352
Battery Holder
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Ideal for studying electricity.
  • Allows you to connect up to 4 D cell batteries in series or parallel (comes configured in series, can be converted though - may require additional wires)
  • Perfect for electronics, physics, or even biology experiments
  • Batteries and cables not included
  • Variable battery power supply
The variable power supply that hold up to 4 D cells is a great tool for use when studying electricity (Batteries and cables not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty