Would you like to log in again?
Demonstration Transformer
Supplier: EISCO SCIENTIFIC LLC
PH0976B
470230-356EA 89 USD InStock
470230-356
Demonstration Transformer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Displays concept of transformers.
  • Step Down Transformer Demonstration with wall plug
  • Main input is 120 V AC, 60 Hz, Output is 4, 6, 12, 24 Volts at 1 Amp
  • Output connectors are standard 4 mm sockets
  • There is a power indicator on the base of the unit
This step down transformer is great for displaying the concept of transformers, or a useful bench apparatus for supplying a variety of voltages using a small foot print and low cost.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty