Would you like to log in again?
Resistance Substitution Board
Supplier: EISCO SCIENTIFIC LLC
PH0992
470230-364EA 38.8 USD InStock
470230-364
Resistance Substitution Board
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Resistor board for electrical circuits.
  • Ideal for circuit testing or electronics classroom investigation
  • 12 Different Resistors selectable via rotary knob
  • Connect to circuit via 4 mm leads
  • 33 ohms to 10 M ohms
This resistor board is extremely useful for all circuit applications. Great for electronics classrooms or labs, or for use in circuits under design or in testing conditions. Comprised of 12 different resistors situated on a board, with standard preferred values from 33 ohms to 10M ohms in step ratio of 1:3. Select the resistance via the rotary switch and connect into a circuit by the 4 mm sockets terminals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty