Would you like to log in again?
Buzzer
Supplier: EISCO SCIENTIFIC LLC
PH1056H
470230-386EA 24.4 USD InStock
470230-386
Buzzer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Simple circuit module.
  • Simple Circuit module can be used with Worcester Circuit board
  • Has two threaded 4 mm terminals with nut to securely hole connectors.
  • Can be used with Worcester Circuit Board Kit ( PH1301)
  • Low Current buzzer approx. 2mA
Simple Circuit module can be used with Worcester Circuit board, Low Current buzzer approx. 2mA. Can be used with Worcester Circuit Board Kit ( PH1301), Has two 4 mm terminals. 2.875" x 2.875" Board. Equipped with zip tie holders on the back of the board.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty