Would you like to log in again?
Heat Transfer Kit, Class Pack of 15
Supplier: EISCO SCIENTIFIC LLC
PH1120144PK15
470230-396KT 403.65 USD InStock
470230-396
Heat Transfer Kit, Class Pack of 15
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Kits for demonstrating thermal energy and heat transfer.
  • Low range thermometer range: –40...50°C
  • High range thermometer range: –30...110°C
  • 15 kits included
Each of the 15 heat transfer kits contains all the items needed (2 Styrofoam calorimeters, 1 aluminum heat transfer bar, low range thermometer, high range thermometer, and apparatus guide) for demonstrating and learning about thermal energy and heat transfer. This product is a great way to introduce students to the concepts of thermal energy and heat transfer in a visually and kinesthetically effective manner.
Ordering information: Each of the 15 kits contains 2 Styrofoam calorimeters with lids, 1 aluminum heat transfer bar, 1 low range thermometer (Plastic), and 1 high range thermometer (metal).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty