Would you like to log in again?
Friction Cube with Four Surfaces
Supplier: EISCO SCIENTIFIC LLC
PH1120151
470230-400EA 18.55 USD InStock
470230-400
Friction Cube with Four Surfaces
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Ideal for exploring force of friction.
  • 2 inch cube with hook
  • Surfaces: Wood, Paper, Vinyl, Sandpaper
  • By pulling the cube by the attached hook, one can both qualitatively and quantitatively demonstrate the properties of static and kinetic friction
The friction cube provides four surfaces for exploration into the force of friction. Each side of the 2" cube provides a surface (wood, paper, vinyl, sandpaper) with a different coefficient of friction. By pulling the cube by the attached hook, one can both qualitatively and quantitatively demonstrate the properties of static and kinetic friction.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty