Would you like to log in again?
Rheostats
Supplier: EISCO SCIENTIFIC LLC
PH1150D PH1150K PH1150H PH1150A PH1150L PH1150G PH1150F PH1150E PH1150J PH1150C
470230-428EA 89.55 USD InStock
470230-428 470230-416 470230-426 470230-414 470230-424 470230-412 470230-422 470230-420 470230-430 470230-418
Rheostats
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Standard resistance rheostat.
  • Rheostat is wound with heavily oxidized constantan (Copper-Nickel Alloy) wire on a vitreous enameled steel tube
  • Heavy duty sliding contact made of phosphor bronze and nickel plated for corrosion resistance
  • Two 4 mm terminal posts which are fitted to facilitate external connection conventional 4 mm plug
This Rheostat is wound with heavily oxidized constantan (Copper-Nickel Alloy) wire on a vitreous enameled steel tube. The tube is supported on die cast ends. There is a heavy duty sliding contact made of phosphor bronze and nickel plated for corrosion resistance. There are two 4 mm terminal posts which are fitted to facilitate external connection conventional 4 mm plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty