Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Demonstration Dynamo

Supplier: EISCO SCIENTIFIC LLC
PH1246
470230-436EA 63.05 USD InStock
470230-436
Demonstration Dynamo
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Hand and motor powered dynamo.

  • Electrical output via 4 mm terminals
  • Hand powered
  • May also be a motor powered by 6 to 8V power supply

This model is mounted on a base plate 200 x 125 mm approx. which also carries a hand-drive pulley 125 mm diameter coupled to the smaller dynamo pulley by a rubber belt to give a step-up ratio. Electrical output is via a pair of 4 mm sockets and a light emitting diode is provided as simple output indicator. The model may also be used as a motor on 6 - 8 V DC supply.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty