Would you like to log in again?
Solar Panel Demonstration, Eisco Scientific
Supplier: Eisco Scientific
PH1320A
470230-442EA 53.3 USD InStock
470230-442
Solar Panel Demonstration, Eisco Scientific
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Compact demonstration of alternative energy.
  • Six solar cells, connected in series on clear base
  • 12 V output, 130 mA current
  • Plugs of 4 mm
Designed for use in experiments related to the transfer of energy and is an excellent teaching tool to introduce students to the fundamentals of one of the most critical energy systems of our future.
The unit itself measures 8.5×6×1.5" and each solar cell measures 3×1". Designed with two 1.4" jacks.
Ordering information: Requires 4 mm leads, not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty