Would you like to log in again?
Solar Cell
Supplier: EISCO SCIENTIFIC LLC
PH1320EIS
470230-444EA 41 USD InStock
470230-444
Solar Cell
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Generate electricity with a solar panel.
  • Selenium voltaic solar cell mounted on a metal base
  • Accepts two 4 mm plug type cords
Demonstrate the conversion between light energy and electricity. Connect a motor via cords to the solar cell and introduce a light source to allow the solar cell to power the motor.
Ordering information: Cords not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty