Would you like to log in again?
Digital to Analogue Convertor
Supplier: EISCO SCIENTIFIC LLC
PH1367
470230-450EA 121.35 USD InStock
470230-450
Digital to Analogue Convertor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrates basis of digital communications.
  • Converts digital signal to an analog output
  • Great for demonstrating the basis of digital communications
  • Companion apparatus to the labs analog to digital converter
This is great companion unit to the Labs Analog to Digital Converter (PH1366). A 3 bit binary input (0 V represents 0 and 5 V represents 1) is converted to 0 to 7 V analogue output. By using the two units together the sampling process can be demonstrated to show that information is lost during the conversion process. The Digital to Analog Converter can be powered from the ADC or from a separate 12 V power supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty