Would you like to log in again?
Visual Scientifics Inclined Plane, 1.5 m
Supplier: EISCO SCIENTIFIC LLC
PTLFIP
470230-454EA 115.9 USD InStock
470230-454
Visual Scientifics Inclined Plane, 1.5 m
Educational Materials Physics Educational Materials Mechanics Learning Activities
This is an extra long version of the Visual Scientifics Inclined Plane, and includes all of the same accessories.
  • Lab manual included, with activities and extensions
  • Covers NGSS and common core standards
  • Platform neutral - works with your preferred probeware/video analysis tools
This inclined plane system makes it simple to demonstrate the relationships between mass and speed, kinetic and potential energy, dynamic equilibrium, work-energy theorem and unbalanced forces. The included lab manual provides approximately 3 hours of laboratory activities. Instructions are written to explicitly cover Next Generation Science Standards and common core standards.
Ordering information: Visual Scientifics Base (470148-822) required, but sold separately.
Delivery information: This kit includes an inclined plane track, sliding protractor and bob, car, pulley attachment, nylon cord, and mounting rods with hardware. Recommended accessories: video camera and graphical analysis software.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty